Learn More
Electron Microscopy Sciences SEM Specimen Mount Mount Forcep
SDP
Catalog No. 5033089
Encompass_Preferred
Made from stainless steel with a serrated handle and a guide pin. The tip is bent at a 45 degree and formed into a ring, which has a diameter of 10 mm when fully closed. 150 mm long.
Catalog No. 50-330-89 Supplier Electron Microscopy Sciences Supplier No. 76805
Made from stainless steel with a serrated handle and a guide pin. The tip is bent at a 45 degree and formed into a ring, which has a diameter of 10 mm when fully closed. 150 mm long.