Learn More
Electron Microscopy Sciences Hitachi SEM Cylinder Mount 9/16", 3/8" height, Aluminum
SDP
Catalog No. 5032988
Encompass_Preferred
Hitachi SEM mounts are made from lathe finished ultra-pure aluminum.
Catalog No. 50-329-88 Supplier Electron Microscopy Sciences Supplier No. 75652
Hitachi SEM mounts are made from lathe finished ultra-pure aluminum.