Learn More
Electron Microscopy Sciences SEM Mount Hitachi M4 1" Plain Surface, 1/4" height, Aluminum
SDP
Catalog No. 5032981
Encompass_Preferred
Hitachi cylinder mount has a M4 tapered hole with female thread. Made from lathe finished ultra-pure aluminum. M4 tapered hole.
Head: 1"
Height: 6 mm
Catalog No. 50-329-81 Supplier Electron Microscopy Sciences Supplier No. 75623
Hitachi cylinder mount has a M4 tapered hole with female thread. Made from lathe finished ultra-pure aluminum. M4 tapered hole.
Head: 1"
Height: 6 mm