Learn More
Electron Microscopy Sciences SEM Mount Hitachi M4 1" Plain Surface, 1/4" height, Aluminum
SDP
Supplier:  Electron Microscopy Sciences 75621
Encompass_Preferred
Hitachi cylinder mount has a M4 tapered hole with female thread. Made from lathe finished ultra-pure aluminum. M4 tapered hole.
Head: 1"
Height: 6 mm
Catalog No. 50-329-83