Learn More
Electron Microscopy Sciences SEM Mount Hitachi M4 1" Plain Surface, 1/4" height, Aluminum
SDP
Catalog No. 5032982
Encompass_Preferred
Hitachi cylinder mount has a M4 tapered hole with female thread. Made from lathe finished ultra-pure aluminum. M4 tapered hole.
Head: 1"
Height: 6 mm
Catalog No. 50-329-82 Supplier Electron Microscopy Sciences Supplier No. 75622
Hitachi cylinder mount has a M4 tapered hole with female thread. Made from lathe finished ultra-pure aluminum. M4 tapered hole.
Head: 1"
Height: 6 mm