Learn More
Electron Microscopy Sciences SEM Hitachi S-450 M4 Mount Plain Surface 5/8",1/4" height, Carbon
SDP
Supplier:  Electron Microscopy Sciences 76470
Encompass_Preferred
Hitachi S-450 SEM mounts with M4 tapered hole and female thread. Made from spectroscopically pure carbon.
Head: 5/8"
Height: 1/4"
Catalog No. 50-329-94