Learn More
Electron Microscopy Sciences SEM Mount Hitachi S-500 Mount Hitachi S-500, 25 mm, Aluminum
SDP
Catalog No. 5032996
Encompass_Preferred
Hitachi S-500 threaded pin SEM mounts are made from lathe finished ultra-pure aluminum.
Head: 1"
Height: 1/4"
Catalog No. 50-329-96 Supplier Electron Microscopy Sciences Supplier No. 75642
Add to Cart
Add to Cart
Hitachi S-500 threaded pin SEM mounts are made from lathe finished ultra-pure aluminum.
Head: 1"
Height: 1/4"