Learn More
Electron Microscopy Sciences SEM Hitachi S-450 M4 Mount 3-Division 5/8", 1/4" height, Aluminum
SDP
Catalog No. 501901642
Encompass_Preferred
Hitachi S-450 SEM mounts with M4 tapered hole and female thread. With 3 divisions. Made from ultra-pure aluminum with standard lathe finish.
Head: 5/8"
Height: 1/4"
Catalog No. 50-190-1642 Supplier Electron Microscopy Sciences Supplier No. 75610D
Hitachi S-450 SEM mounts with M4 tapered hole and female thread. With 3 divisions. Made from ultra-pure aluminum with standard lathe finish.
Head: 5/8"
Height: 1/4"