Learn More
Electron Microscopy Sciences SEM Mount Holder Box for 3.2 mm Pins
SDP
Catalog No. 5028822
Encompass_Preferred
SEM specimen mount holder box with insert and lid holds 12 mounts.
Use: 12.5 mm head and 1/8" diameter
Catalog No. 50-288-22 Supplier Electron Microscopy Sciences Supplier No. 76620
SEM specimen mount holder box with insert and lid holds 12 mounts.
Use: 12.5 mm head and 1/8" diameter