Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences SEM Mount Holder Box for 3.2 mm Pins
SDP

Catalog No. 5028823
Encompass_Preferred

SEM specimen mount holder box with insert and lid holds 4 pin mounts
Use: 12.5 mm head and 1/8" diameter

Catalog No. 50-288-23 Supplier Electron Microscopy Sciences Supplier No. 76600
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SEM specimen mount holder box with insert and lid holds 4 pin mounts
Use: 12.5 mm head and 1/8" diameter

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.