Learn More
Electron Microscopy Sciences SEM Mount Holder Box for JEOL
SDP
Catalog No. 5028827
Encompass_Preferred
Box with insert and lid.
Use: 10 mm x 10 mm cylinders
Catalog No. 50-288-27 Supplier Electron Microscopy Sciences Supplier No. 76710
Box with insert and lid.
Use: 10 mm x 10 mm cylinders