Learn More
Electron Microscopy Sciences SEM Mount Holder Box for JEOL 840
SDP
Supplier:  Electron Microscopy Sciences 76732
Encompass_Preferred
SEM specimen mount holder box with insert and lid holds 4 mounts for JEOL 840.
Use: 12.5 mm x 12.5 mm cylinders
Catalog No. 50-288-30