Learn More
Electron Microscopy Sciences SEM Mount Holder Box for JEOL 840
SDP
Catalog No. 5028829
Encompass_Preferred
SEM specimen mount holder box with insert and lid holds 4 mounts for JEOL 840.
Use: 12.5 mm x 12.5 mm cylinders
Catalog No. 50-288-29 Supplier Electron Microscopy Sciences Supplier No. 76734
SEM specimen mount holder box with insert and lid holds 4 mounts for JEOL 840.
Use: 12.5 mm x 12.5 mm cylinders