Learn More
Electron Microscopy Sciences Hitachi M4 SEM Cylinder Mount 1-1/4" surface, 1/4" height, Aluminum
SDP
Catalog No. 501901471
Encompass_Preferred
Hitachi M4 SEM mounts are made from lathe finished ultra-pure aluminum.
Head: 1-1/4
Pin height: 1/4"
Catalog No. 50-190-1471 Supplier Electron Microscopy Sciences Supplier No. 75660
Hitachi M4 SEM mounts are made from lathe finished ultra-pure aluminum.
Head: 1-1/4
Pin height: 1/4"