Learn More
Electron Microscopy Sciences ISI, ABT, Topcon SEM Cylinder Mount 23.7 mm surface, 9.5 mm height, Aluminum
SDP
Supplier:  Electron Microscopy Sciences 75437
Encompass_Preferred
ISI, ABT, Topcon specimen mounts are made from ultra-pure aluminum, lathe finish.
Head: 23.7 mm
Height: 9.5 mm
Catalog No. 50-330-01