Learn More
Electron Microscopy Sciences JEOL SEM Cylinder Mount 1" surface, 8 mm height, Aluminum
SDP
Catalog No. 501928165
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1"
Height: 8 mm
Catalog No. 50-192-8165 Supplier Electron Microscopy Sciences Supplier No. 75693
Add to Cart
Add to Cart
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1"
Height: 8 mm