Learn More
Electron Microscopy Sciences JEOL SEM Cylinder Mount 1" surface, 8 mm height, Aluminum
SDP
Catalog No. 501928167
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1"
Height: 8 mm
Catalog No. 50-192-8167 Supplier Electron Microscopy Sciences Supplier No. 75695
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1"
Height: 8 mm