Learn More
Electron Microscopy Sciences JEOL SEM Cylinder Mount 1-1/4" surface, 13/16" height, Aluminum
SDP
Catalog No. 501928171
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1-1/4"
Height: 13/16"
Catalog No. 50-192-8171 Supplier Electron Microscopy Sciences Supplier No. 7583076
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1-1/4"
Height: 13/16"