Learn More
Electron Microscopy Sciences JEOL SEM Cylinder Mount 2" surface 3- Division, 3/8" height, Aluminum
SDP
Catalog No. 501927102
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 2"
Height: 3/8"
Catalog No. 50-192-7102 Supplier Electron Microscopy Sciences Supplier No. 75708
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 2"
Height: 3/8"