Learn More
Electron Microscopy Sciences JEOL SEM Cylinder Mount 1" surface, 1/2" height, Aluminum
SDP
Catalog No. 5033003
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1"
Height: 1/2"
Catalog No. 50-330-03 Supplier Electron Microscopy Sciences Supplier No. 75690
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1"
Height: 1/2"