Learn More
Electron Microscopy Sciences Cylinder Mount SEM 3/8" head, 3/16" height, Aluminum
SDP
Catalog No. 5026750
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 3/8"
Pin height: 3/16"
Catalog No. 50-267-50 Supplier Electron Microscopy Sciences Supplier No. 75350
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 3/8"
Pin height: 3/16"