Learn More
Electron Microscopy Sciences JEOL SEM Cylinder Mount 10 mm Head, 10 mm height, Aluminum
SDP
Catalog No. 5033036
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with standard lathe finish.
Head: 3/8"
Height: 3/8"
Catalog No. 50-330-36 Supplier Electron Microscopy Sciences Supplier No. 75320
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with standard lathe finish.
Head: 3/8"
Height: 3/8"