Learn More
Electron Microscopy Sciences JEOL SEM Cylinder Mount 1/2" surface, 3/8" height, Aluminum
SDP
Catalog No. 5033019
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL JSM-840 scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1/2"
Height: 3/8"
Catalog No. 50-330-19 Supplier Electron Microscopy Sciences Supplier No. 75732
Cylinder specimen mounts are for use with JEOL JSM-840 scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1/2"
Height: 3/8"