Learn More
Electron Microscopy Sciences JEOL SEM Cylinder Mount 10 mm Head, 10 mm height, Carbon
SDP
Supplier:  Electron Microscopy Sciences 76320
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from spectroscopically pure carbon.
Head: 3/8"
Height: 3/8"
Catalog No. 50-330-42