Learn More
Electron Microscopy Sciences JEOL ISI SEM Cylinder Mount 5/8" surface, 5/8" height, Aluminum
SDP
Supplier:  Electron Microscopy Sciences 75410
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL and ISI scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 5/8"
Height: 5/8"
Catalog No. 50-330-16