Learn More
Electron Microscopy Sciences JEOL ISI SEM Cylinder Mount 5/8" surface, 3/8" height, Aluminum
SDP
Catalog No. 5033012
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL and ISI scanning electron microscopes. The mounts are made from ultra-pure aluminum with standard lathe finish.
Head: 5/8"
Height: 3/8"
Catalog No. 50-330-12 Supplier Electron Microscopy Sciences Supplier No. 75460
Cylinder specimen mounts are for use with JEOL and ISI scanning electron microscopes. The mounts are made from ultra-pure aluminum with standard lathe finish.
Head: 5/8"
Height: 3/8"