Learn More
Electron Microscopy Sciences JEOL ISI SEM Cylinder Mount 5/8" surface, 3/8" height, Carbon
SDP
Supplier:  Electron Microscopy Sciences 76440
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL and ISI scanning electron microscopes. The mounts are made from spectroscopically pure carbon.
Head: 5/8"
Height: 3/8"
Catalog No. 50-330-10