Learn More
Electron Microscopy Sciences Cylinder Mount JEOL ISI ABT Topcon SEM 9.5 mm Head, 9.5 mm height, Aluminum
SDP
Catalog No. 501928161
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL, ISI, ABT, and Topcon scanning electron microscopes. Made from ultra-pure aluminum with a standard lathe finish.
Head: 9.5 mm
Height: 9.5 mm
Catalog No. 50-192-8161 Supplier Electron Microscopy Sciences Supplier No. 75258
Cylinder specimen mounts are for use with JEOL, ISI, ABT, and Topcon scanning electron microscopes. Made from ultra-pure aluminum with a standard lathe finish.
Head: 9.5 mm
Height: 9.5 mm