Learn More
Electron Microscopy Sciences JEOL SEM JSM 840 Cylinder Mount 1" surface, 1/4" height, Aluminum
SDP
Supplier:  Electron Microscopy Sciences 75700
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL JSM-840 scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1"
Height: 1/4"
Catalog No. 50-330-27