Learn More
Electron Microscopy Sciences JEOL JSM 840 SEM Cylinder Mount 1/2" surface, 1/4" height, Carbon
SDP
Catalog No. 5033025
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL JSM-840 scanning electron microscopes. The mounts are made from spectroscopically pure carbon.
Head: 1/2"
Height: 1/4"
Catalog No. 50-330-25 Supplier Electron Microscopy Sciences Supplier No. 76330
Cylinder specimen mounts are for use with JEOL JSM-840 scanning electron microscopes. The mounts are made from spectroscopically pure carbon.
Head: 1/2"
Height: 1/4"