Learn More
Electron Microscopy Sciences Shorter Pin SEM Stub Slotted 1/2" Head with Shorter Pin (5 mm), Aluminum, 3.1 mm Pin diameter
SDP
Catalog No. 5024452
Encompass_Preferred
SEM stub specimen mount with tapered end pin and slotted head for use with LEO SEMs. Made from ultra-pure aluminum with a standard lathe finish.
Head: 1/2"
Pin height: 5 mm
Catalog No. 50-244-52 Supplier Electron Microscopy Sciences Supplier No. 75190
SEM stub specimen mount with tapered end pin and slotted head for use with LEO SEMs. Made from ultra-pure aluminum with a standard lathe finish.
Head: 1/2"
Pin height: 5 mm