Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Large slotted 1-1/4" head with 1/2" Pin, Aluminum, 1/8" Pin diameter
SDP
Catalog No. 5032979
Encompass_Preferred
Large slotted head pin stub mount are for Leica, Cambridge, LEO, FEI, Phillips, Zeiss, and Camscan SEMs specimen mounts. Made from lathe finished ultra-pure aluminum.
Head: 1-1/4"
Pin height: 1/2"
Pin diameter: 1/8"
Catalog No. 50-329-79 Supplier Electron Microscopy Sciences Supplier No. 75187
Large slotted head pin stub mount are for Leica, Cambridge, LEO, FEI, Phillips, Zeiss, and Camscan SEMs specimen mounts. Made from lathe finished ultra-pure aluminum.
Head: 1-1/4"
Pin height: 1/2"
Pin diameter: 1/8"