Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Large slotted 1-1/4" head with 1/2" Pin, Aluminum, 1/8" Pin diameter
SDP
Supplier:  Electron Microscopy Sciences 75188
Encompass_Preferred
Large slotted head pin stub mount are for Leica, Cambridge, LEO, FEI, Phillips, Zeiss, and Camscan SEMs specimen mounts. Made from lathe finished ultra-pure aluminum.
Head: 1-1/4"
Pin height: 1/2"
Pin diameter: 1/8"
Catalog No. 50-329-78