Learn More
Electron Microscopy Sciences Shorter Pin Mini SEM Stub Mini Pin Stub with Short Pin, Aluminum
SDP
Catalog No. 501928194
Encompass_Preferred
Mini SEM stub specimen mount with shorter pin (6 mm) for use with all Zeiss/LEO scanning electron microscopes, FE-SEMs & FIB systems. Made from ultra-pure aluminum with a standard lathe finish.
Head: 6.6 x 1.3 mm
Pin height: 6.25 mm
Catalog No. 50-192-8194 Supplier Electron Microscopy Sciences Supplier No. 75824
Mini SEM stub specimen mount with shorter pin (6 mm) for use with all Zeiss/LEO scanning electron microscopes, FE-SEMs & FIB systems. Made from ultra-pure aluminum with a standard lathe finish.
Head: 6.6 x 1.3 mm
Pin height: 6.25 mm