Learn More
Electron Microscopy Sciences Shorter Pin Mini SEM Stub Mini Pin Stub with Short Pin, Stainless
SDP
Catalog No. 501928198
Encompass_Preferred
Mini SEM stub specimen mount with shorter pin (6 mm) for use with all Zeiss/LEO scanning electron microscopes, FE-SEMs & FIB systems. Made of stainless steel.
Head: 6.6 x 1.3 mm
Pin height: 6.25 mm
Catalog No. 50-192-8198 Supplier Electron Microscopy Sciences Supplier No. 75828
Mini SEM stub specimen mount with shorter pin (6 mm) for use with all Zeiss/LEO scanning electron microscopes, FE-SEMs & FIB systems. Made of stainless steel.
Head: 6.6 x 1.3 mm
Pin height: 6.25 mm