Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Non Slotted 1 " Head with 8 mm Pin, Aluminum, 3.1 mm Pin diameter
SDP
Catalog No. 5033753
Encompass_Preferred
Cambridge, Leo, Philips 500, Cameca SEMs specimen mounts made from lathe finished ultra-pure aluminum. Non-slotted head.
Head: 1"
Pin height: 8 mm
Pin diameter: 3.1 mm
Catalog No. 50-337-53 Supplier Electron Microscopy Sciences Supplier No. 75186
Cambridge, Leo, Philips 500, Cameca SEMs specimen mounts made from lathe finished ultra-pure aluminum. Non-slotted head.
Head: 1"
Pin height: 8 mm
Pin diameter: 3.1 mm