Learn More
Electron Microscopy Sciences SEM Hitachi Mounts Rectangular 11 mm x 5.5 mm, 3.5 mm height
SDP
Catalog No. 501928191
Encompass_Preferred
Rectangular SEM specimen mounts for use with Hitachi S-5200/S-5500/SU-9000. Made from ultra-pure aluminum. Available in 11 mm x 5.5 mm with 2, 3.5, 5 or 5..5 mm height.
Size: 3.5 mm
Catalog No. 50-192-8191 Supplier Electron Microscopy Sciences Supplier No. 7546311
Rectangular SEM specimen mounts for use with Hitachi S-5200/S-5500/SU-9000. Made from ultra-pure aluminum. Available in 11 mm x 5.5 mm with 2, 3.5, 5 or 5..5 mm height.
Size: 3.5 mm