Learn More
Electron Microscopy Sciences Shorter Pin SEM Stub Slotted 2" Head, 6 mm Pin, Aluminum, 3.2 mm Pin diameter
SDP
Catalog No. 501928133
Encompass_Preferred
SEM specimen mount with shorter pin and slotted head for use with all Zeiss/LEO SEM, FESEM & FIB systems. Made from ultra-pure aluminum with a standard lathe finish.
Head: 2"
Pin height: 6 mm
Pin diameter: 3.2 mm
Catalog No. 50-192-8133 Supplier Electron Microscopy Sciences Supplier No. 7563950
SEM specimen mount with shorter pin and slotted head for use with all Zeiss/LEO SEM, FESEM & FIB systems. Made from ultra-pure aluminum with a standard lathe finish.
Head: 2"
Pin height: 6 mm
Pin diameter: 3.2 mm