Learn More
Electron Microscopy Sciences Shorter Pin SEM Stub Slotted 38 mm head with 6 mm Pin, Aluminum, 1/8" Pin diameter
SDP
Supplier:  Electron Microscopy Sciences 7518371
Encompass_Preferred
SEM stub specimen mount with shorter pin (6 mm) and 38 mm slotted head for use with Zeiss/LEO scanning electron microscopes, FE-SEMs & FIB systems. Made from ultra-pure aluminum with a standard lathe finish.
Head: 38 mm
Pin height: 6 mm
Pin diameter: 1/8"
Catalog No. 50-192-8152