Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Slotted 1/2" Head with 8 mm Pin, Aluminum, 1/8" Pin diameter
SDP
Supplier:  Electron Microscopy Sciences 75236
Encompass_Preferred
Slotted head pin stub mounts with tappered pin ends are for use with Cambridge, Phillips, Camscan, B&L, Etec, Zeiss. Made from ultra-pure aluminum polished finish with a smooth and luster surface.
Head: 1/2"
Pin height: 8 mm
Pin diameter: 1/8"
Catalog No. 50-248-51