Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Slotted 1/2" Head with 8 mm Pin, Aluminum, 1/8" Pin diameter
SDP
Catalog No. 5024850
Encompass
Slotted head pin stub mounts with tappered pin ends are for use with Cambridge, Phillips, Camscan, B&L, Etec, Zeiss. Made from ultra-pure aluminum polished finish with a smooth and luster surface.
Head: 1/2"
Pin height: 8 mm
Pin diameter: 1/8"
Catalog No. 50-248-50 Supplier Electron Microscopy Sciences Supplier No. 75238
Slotted head pin stub mounts with tappered pin ends are for use with Cambridge, Phillips, Camscan, B&L, Etec, Zeiss. Made from ultra-pure aluminum polished finish with a smooth and luster surface.
Head: 1/2"
Pin height: 8 mm
Pin diameter: 1/8"