Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Slotted 38 mm head with 8 mm Pin, Aluminum, 3.2 mm Pin diameter
SDP
Catalog No. 501928149
Encompass_Preferred
Pin stub mount with slotted head for use with Leica, Cambridge, LEO, FEI, Phillips, PHENOM, Zeiss, Camscan. Made from ultra-pure aluminum with a standard lathe finish.
Head: 38 mm
Pin height: 8 mm
Pin diameter: 3.2 mm
Catalog No. 50-192-8149 Supplier Electron Microscopy Sciences Supplier No. 7518367
Pin stub mount with slotted head for use with Leica, Cambridge, LEO, FEI, Phillips, PHENOM, Zeiss, Camscan. Made from ultra-pure aluminum with a standard lathe finish.
Head: 38 mm
Pin height: 8 mm
Pin diameter: 3.2 mm