Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Slotted 1" head with 1/2" Pin, Aluminum, 3.1 mm Pin diameter
SDP
Catalog No. 5032975
Encompass_Preferred
Scanning electron microscope specimen mounts for FEI/Philips, ZEISS/LEO, Cambridge, Leica, Amray, Tescan and Camscan SEMs. Made from lathe finished ultra-pure aluminum.
Head: 1"
Pin height: 1/2"
Pin diameter: 3.1 mm
Catalog No. 50-329-75 Supplier Electron Microscopy Sciences Supplier No. 75184
Scanning electron microscope specimen mounts for FEI/Philips, ZEISS/LEO, Cambridge, Leica, Amray, Tescan and Camscan SEMs. Made from lathe finished ultra-pure aluminum.
Head: 1"
Pin height: 1/2"
Pin diameter: 3.1 mm