Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Slotted 3/8" head with 7/16" Pin, Aluminum, 3.1 mm Pin diameter
SDP
Catalog No. 5032974
Encompass_Preferred
Small, slotted head pin stub mounts are for Cambridge, LEO, Philips 500, Cameca SEMs. Made from lathe finished ultra-pure aluminum.
Head: 3/8"
Height: 7/16"
Pin diameter: 3.1 mm
Catalog No. 50-329-74 Supplier Electron Microscopy Sciences Supplier No. 75181
Small, slotted head pin stub mounts are for Cambridge, LEO, Philips 500, Cameca SEMs. Made from lathe finished ultra-pure aluminum.
Head: 3/8"
Height: 7/16"
Pin diameter: 3.1 mm