Learn More
Electron Microscopy Sciences SEM Mount Storage Tube
SDP
Catalog No. 5028588
Encompass_Preferred
A plastic tube and plug for storing SEM pin stub mounts. The pin mount 1/8" (3.1mm) in diameter is inserted securely to the plug and protected by the tube. Ideal for storage or mailing.
Catalog No. 50-285-88 Supplier Electron Microscopy Sciences Supplier No. 7653001
A plastic tube and plug for storing SEM pin stub mounts. The pin mount 1/8" (3.1mm) in diameter is inserted securely to the plug and protected by the tube. Ideal for storage or mailing.