Learn More
Electron Microscopy Sciences SEM Single Mount Storage Tube 100/pk
SDP
Catalog No. 5028587
Encompass_Preferred
A plastic tube and plug for storing SEM pin stub mounts. The pin mount 1/8" (3.1mm) in diameter is inserted securely to the plug and protected by the tube. Ideal for storage or mailing.
Catalog No. 50-285-87 Supplier Electron Microscopy Sciences Supplier No. 7653010
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A plastic tube and plug for storing SEM pin stub mounts. The pin mount 1/8" (3.1mm) in diameter is inserted securely to the plug and protected by the tube. Ideal for storage or mailing.