Learn More
Electron Microscopy Sciences SEM Specimen Mount Mount Tweezers Style 2E1/8
SDP
Catalog No. 5028833
Encompass_Preferred
Dumont Specimen mount tweezers are the finest tweezers available. Choose the one that best suits your needs.
Dumoxel is composed of C, Cr, Ni, Mo and Cu and offers the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel is 95% antimagnetic, resistant up to temperatures of around 400 degree C (DIN 50 914) and is suitable for autoclave sterilization at 270 degree C.
Size: 0=3.2 mm
Catalog No. 50-288-33 Supplier Electron Microscopy Sciences Supplier No. 76804
Dumont Specimen mount tweezers are the finest tweezers available. Choose the one that best suits your needs.
Dumoxel is composed of C, Cr, Ni, Mo and Cu and offers the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel is 95% antimagnetic, resistant up to temperatures of around 400 degree C (DIN 50 914) and is suitable for autoclave sterilization at 270 degree C.
Size: 0=3.2 mm