Learn More
Electron Microscopy Sciences SEM Pin Stub Zeiss Flat End Pin, Slotted 1/2" head, Aluminum
SDP
Catalog No. 5033045
Encompass_Preferred
Zeiss SEM mounts. Flat end pin and slotted head. Made from ultra-pure aluminum with standard lathe finish.
Head: 1/2"
Height: 1/8"
Catalog No. 50-330-45 Supplier Electron Microscopy Sciences Supplier No. 75510
Zeiss SEM mounts. Flat end pin and slotted head. Made from ultra-pure aluminum with standard lathe finish.
Head: 1/2"
Height: 1/8"