Learn More
Electron Microscopy Sciences SEM Paper Storage Box
SDP
Catalog No. 5034404
Encompass_Preferred
An inexpensive solution for storing Pin Mounts with a Pop Up box which stores up to 8 mounts with 1/2" head and 1/8" diameter pin. Box Dimensions: 7cm long x 3.8cm wide x 3.5cm high.
Catalog No. 50-344-04 Supplier Electron Microscopy Sciences Supplier No. 76505
An inexpensive solution for storing Pin Mounts with a Pop Up box which stores up to 8 mounts with 1/2" head and 1/8" diameter pin. Box Dimensions: 7cm long x 3.8cm wide x 3.5cm high.