Learn More
Electron Microscopy Sciences SEM Pin Stub Adapter, 15 mm
SDP
Catalog No. 501930523
Encompass_Preferred
A simple to use adapter which will convert Hitachi, JEOL or ISI microscopes to use 1/8" pin stubs or Cambridge stubs. Made of aluminum with spring retainers which will give a good connection.
Catalog No. 50-193-0523 Supplier Electron Microscopy Sciences Supplier No. EMS061
A simple to use adapter which will convert Hitachi, JEOL or ISI microscopes to use 1/8" pin stubs or Cambridge stubs. Made of aluminum with spring retainers which will give a good connection.